Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcoln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053177.1 | NP_444407.1 | 10 - 11 | 1307 | 66 | |
GenBank mRNA | AF302009.1 | - | 10 - 11 | 1304 | 66 | |
AK028385.1 | - | 10 - 11 | 1284 | 66 | ||
AK034471.1 | - | 10 - 11 | 1318 | 66 | ||
AK049606.1 | - | 10 - 11 | 1308 | 66 | ||
BC005651.1 | - | 10 - 11 | 1307 | 3257440 | 66 | |
BC020377.1 | - | 6 - 7 | 1749 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |