Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Colq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009937.2 | NP_034067.2 | 12 - 13 | 955 | 85 | |
GenBank mRNA | AK134212.1 | - | 12 - 13 | 959 | 85 | |
BC107386.2 | - | 12 - 13 | 889 | 40057706 | 85 | |
BC107387.1 | - | 11 - 12 | 752 | 40057715 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |