Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdr16c5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181989.1 | NP_871789.1 | 2 - 3 | 554 | 80 | |
GenBank mRNA | AK136508.1 | - | 1 - 2 | 525 | 80 | |
AY277588.1 | - | 2 - 3 | 554 | 80 | ||
BC104134.1 | - | 2 - 3 | 473 | 40045268 | 80 | |
BC104135.1 | - | 2 - 3 | 473 | 40045269 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |