Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400019249.mm.1 |
NM_001012450.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019249.mm.1 |
NM_001012450.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019249.mm.1 |
NM_080471.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019249.mm.1 |
NM_080471.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019249.mm.1 |
TC0400002367.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400019248.mm.1 |
NM_001012450.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019248.mm.1 |
NM_001012450.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019248.mm.1 |
NM_080471.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019248.mm.1 |
NM_080471.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019248.mm.1 |
TC0400002367.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400019242.mm.1 |
NM_001012450.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019242.mm.1 |
NM_001012450.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019242.mm.1 |
NM_001012451.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019242.mm.1 |
NM_001012451.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019242.mm.1 |
NM_080471.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019242.mm.1 |
NM_080471.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019242.mm.1 |
TC0400002367.mm.1 |
MTA TC |