Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016979.1 | NP_058675.1 | 7 - 8 | 1198 | 66 | |
GenBank mRNA | AJ238004.1 | - | 7 - 8 | 1081 | 66 | |
AK034681.1 | - | 7 - 8 | 1517 | 66 | ||
AK037141.1 | - | 7 - 8 | 1192 | 66 | ||
AK081548.1 | - | 7 - 8 | 1178 | 66 | ||
AK139510.1 | - | 7 - 8 | 1206 | 66 | ||
AK149937.1 | - | 7 - 8 | 1176 | 66 | ||
AK150588.1 | - | 7 - 8 | 1160 | 66 | ||
AK152166.1 | - | 7 - 8 | 994 | 66 | ||
AK154447.1 | - | 7 - 8 | 1178 | 66 | ||
AK169322.1 | - | 7 - 8 | 1149 | 66 | ||
BC006875.1 | - | 7 - 8 | 1198 | 3596013 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |