Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199042.2 | NP_950243.1 | 1 - 2 | 353 | 80 | |
GenBank mRNA | BC038639.1 | - | 1 - 2 | 309 | 5118383 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |