Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gon4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242372.1 | NP_001229301.1 | 5419 | 58 | ||
NM_027389.3 | NP_081665.2 | 5368 | 58 | |||
GenBank mRNA | AK122531.1 | - | 4740 | 58 | ||
AK147575.1 | - | 5370 | 58 | |||
AK166365.1 | - | 390 | 58 | |||
BC016616.1 | - | 740 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |