Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfmbt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166531.1 | NP_001160003.1 | 18 - 19 | 2175 | 107 | |
NM_001166532.1 | NP_001160004.1 | 17 - 18 | 2269 | 107 | ||
NM_019460.2 | NP_062333.1 | 17 - 18 | 2126 | 107 | ||
GenBank mRNA | AB032165.1 | - | 16 - 17 | 1971 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |