Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028744.3 | NP_083020.2 | 1 - 2 | 395 | 154 | |
GenBank mRNA | AK016754.1 | - | 1 - 2 | 361 | 154 | |
AY148879.1 | - | 361 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |