Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400012840.mm.1 |
NM_022656.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012840.mm.1 |
NM_022656.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012840.mm.1 |
TC1400001672.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012846.mm.1 |
NM_022656.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012846.mm.1 |
NM_022656.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012846.mm.1 |
TC1400001672.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012848.mm.1 |
NM_022656.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012848.mm.1 |
NM_022656.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012848.mm.1 |
TC1400001672.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012835.mm.1 |
NM_022656.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012835.mm.1 |
NM_022656.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012835.mm.1 |
TC1400001672.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012838.mm.1 |
NM_022656.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012838.mm.1 |
NM_022656.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012838.mm.1 |
TC1400001672.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012855.mm.1 |
NM_022656.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012855.mm.1 |
NM_022656.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012855.mm.1 |
TC1400001672.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012839.mm.1 |
NM_022656.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012839.mm.1 |
NM_022656.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012839.mm.1 |
TC1400001672.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012856.mm.1 |
NM_022656.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012856.mm.1 |
NM_022656.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012856.mm.1 |
TC1400001672.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012842.mm.1 |
NM_022656.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012842.mm.1 |
NM_022656.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012842.mm.1 |
TC1400001672.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012836.mm.1 |
NM_022656.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012836.mm.1 |
NM_022656.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012836.mm.1 |
TC1400001672.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012852.mm.1 |
NM_022656.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012852.mm.1 |
NM_022656.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012852.mm.1 |
TC1400001672.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012853.mm.1 |
NM_022656.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012853.mm.1 |
NM_022656.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012853.mm.1 |
TC1400001672.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012851.mm.1 |
NM_022656.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012851.mm.1 |
NM_022656.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012851.mm.1 |
TC1400001672.mm.1 |
MTA TC |