Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nisch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022656.2 | NP_073147.2 | 13 - 14 | 1688 | 145 | |
GenBank mRNA | AF315344.1 | - | 9 - 10 | 1123 | 145 | |
BC089329.1 | - | 8 - 9 | 907 | 145 | ||
BC108364.1 | - | 8 - 9 | 883 | 145 | ||
BC144980.1 | - | 9 - 10 | 1017 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |