Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dthd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170705.1 | NP_001164176.1 | 4 - 5 | 1330 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |