Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028234.1 | NP_082510.1 | 13 - 14 | 2380 | 65 | |
GenBank mRNA | AK153360.1 | - | 2 - 3 | 476 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |