Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4bp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133898.4 | NP_598659.3 | 1 - 2 | 262 | 148 | |
GenBank mRNA | AK046116.1 | - | 1 - 2 | 238 | 148 | |
AK131647.1 | - | 1 - 2 | 254 | 148 | ||
AK138666.1 | - | 1 - 2 | 238 | 148 | ||
BB629746.1 | - | 1 - 2 | 262 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |