Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK030269.1 | - | 4 - 5 | 581 | 65 | |
AK041560.1 | - | 3 - 4 | 269 | 65 | ||
BC125478.1 | - | 4 - 5 | 566 | 40129993 | 65 | |
BC125480.1 | - | 4 - 5 | 566 | 40129995 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |