Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301424.1 | NP_001288353.1 | 3 - 4 | 541 | 63 | |
NM_007513.4 | NP_031539.3 | 3 - 4 | 657 | 63 | ||
GenBank mRNA | AK139342.1 | - | 3 - 4 | 633 | 63 | |
AK147991.1 | - | 3 - 4 | 566 | 63 | ||
AK163202.1 | - | 3 - 4 | 541 | 63 | ||
BC068234.1 | - | 3 - 4 | 549 | 63 | ||
BC145779.1 | - | 2 - 3 | 462 | 40131082 | 63 | |
BC145781.1 | - | 2 - 3 | 462 | 40131084 | 63 | |
JN226407.1 | - | 1 - 2 | 377 | 63 | ||
JN226408.1 | - | 1 - 2 | 377 | 63 | ||
M26687.1 | - | 3 - 4 | 575 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |