Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183161.3 | NP_898984.3 | 5 - 6 | 687 | 72 | |
GenBank mRNA | CF165170.1 | - | 2 - 3 | 173 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mus musculus Probe spans exons |