Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027338.1 | NP_081614.1 | 2 - 3 | 180 | 75 | |
GenBank mRNA | AK008946.1 | - | 2 - 3 | 218 | 75 | |
AK150754.1 | - | 2 - 3 | 201 | 75 | ||
AK151035.1 | - | 2 - 3 | 201 | 75 | ||
AK156241.1 | - | 194 | 75 | |||
AK161865.1 | - | 232 | 75 | |||
BC010811.1 | - | 2 - 3 | 180 | 4210911 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |