Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177111.3 | NP_796085.3 | 4 - 5 | 567 | 119 | |
GenBank mRNA | AK145297.1 | - | 3 - 4 | 580 | 119 | |
BC070471.1 | - | 4 - 5 | 567 | 30626499 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |