Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500006870.mm.1 |
TC0500000790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500006870.mm.1 |
NM_177680.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500006870.mm.1 |
NM_177680.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500006875.mm.1 |
TC0500000790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500006875.mm.1 |
NM_177680.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500006875.mm.1 |
NM_177680.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500006876.mm.1 |
TC0500000790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500006876.mm.1 |
NM_177680.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500006876.mm.1 |
NM_177680.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500006871.mm.1 |
TC0500000790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500006871.mm.1 |
NM_177680.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500006871.mm.1 |
NM_177680.3 |
Refseq |