Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubqln4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033526.2 | NP_277068.1 | 6 - 7 | 1192 | 64 | |
GenBank mRNA | AB040050.1 | - | 6 - 7 | 1192 | 64 | |
AK150211.1 | - | 6 - 7 | 1129 | 64 | ||
AK156515.1 | - | 6 - 7 | 1146 | 64 | ||
BC017686.1 | - | 6 - 7 | 1154 | 4236887 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |