Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK144138.1 | - | 15 - 16 | 2361 | 67 | |
BC021371.1 | - | 5 - 6 | 604 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |