Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290628.1 | NP_001277557.1 | 7 - 8 | 1335 | 141 | |
NM_139061.5 | NP_620692.3 | 7 - 8 | 1371 | 141 | ||
GenBank mRNA | AF424699.2 | - | 7 - 8 | 1122 | 141 | |
AK036667.1 | - | 1 - 2 | 117 | 141 | ||
AK044995.1 | - | 1 - 2 | 151 | 141 | ||
AK077306.1 | - | 5 - 6 | 753 | 141 | ||
BC025012.2 | - | 7 - 8 | 1046 | 5042879 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |