Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144844.2 | NP_659093.2 | 3 - 4 | 260 | 130 | |
GenBank mRNA | AK148002.1 | - | 3 - 4 | 250 | 130 | |
AK212580.1 | - | 3 - 4 | 164 | 130 | ||
AY046947.1 | - | 3 - 4 | 277 | 130 | ||
BC006915.1 | - | 3 - 4 | 257 | 3601148 | 130 | |
BC049802.1 | - | 3 - 4 | 226 | 6332730 | 130 | |
BY263345.1 | - | 3 - 4 | 260 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |