Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vprbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015507.2 | NP_001015507.1 | 9 - 10 | 1413 | 99 | |
GenBank mRNA | AK122372.1 | - | 9 - 10 | 1376 | 99 | |
BC065119.1 | - | 9 - 10 | 1413 | 6853519 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |