Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008362.2 | NP_032388.1 | 2 - 3 | 291 | 164 | |
GenBank mRNA | BC109135.2 | - | 2 - 3 | 197 | 40039321 | 164 |
M20658.1 | - | 2 - 3 | 291 | 164 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |