Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008513.3 | NP_032539.2 | 12 - 13 | 2949 | 82 | |
GenBank mRNA | AF064984.1 | - | 12 - 13 | 2939 | 82 | |
AF077847.1 | - | 12 - 13 | 2827 | 82 | ||
AK147068.1 | - | 12 - 13 | 2942 | 82 | ||
AK147231.1 | - | 13 - 14 | 3060 | 82 | ||
AK147330.1 | - | 13 - 14 | 3060 | 82 | ||
AK220186.1 | - | 12 - 13 | 2902 | 82 | ||
BC011374.1 | - | 12 - 13 | 2931 | 3154246 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |