Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039692.1 | NP_001034781.1 | 12 - 13 | 2117 | 127 | |
NM_029277.2 | NP_083553.2 | 11 - 12 | 2042 | 127 | ||
GenBank mRNA | AK031648.1 | - | 12 - 13 | 1961 | 127 | |
AK077063.1 | - | 12 - 13 | 2118 | 127 | ||
BC024633.1 | - | 7 - 8 | 529 | 127 | ||
BC037608.1 | - | 11 - 12 | 1857 | 127 | ||
BC141184.1 | - | 11 - 12 | 1851 | 9055796 | 127 | |
BC145506.1 | - | 11 - 12 | 1850 | 9054069 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |