Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK090073.1 | - | 11 - 12 | 1581 | 68 | |
AK148298.1 | - | 11 - 12 | 1578 | 68 | ||
AK173247.1 | - | 12 - 13 | 1818 | 68 | ||
S79463.1 | - | 11 - 12 | 1432 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |