Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asrgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025610.3 | NP_079886.2 | 2 - 3 | 247 | 101 | |
GenBank mRNA | AK006147.1 | - | 2 - 3 | 283 | 101 | |
AK010381.1 | - | 2 - 3 | 289 | 101 | ||
AK030209.1 | - | 2 - 3 | 248 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |