Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172774.3 | NP_766362.2 | 1 - 2 | 367 | 101 | |
GenBank mRNA | AK020102.1 | - | 1 - 2 | 350 | 101 | |
AK031257.1 | - | 1 - 2 | 357 | 101 | ||
AK171423.1 | - | 1 - 2 | 322 | 101 | ||
BB649616.1 | - | 1 - 2 | 368 | 101 | ||
BC078458.1 | - | 1 - 2 | 314 | 6832436 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |