Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080556.3 | NP_542123.3 | 1 - 2 | 349 | 81 | |
GenBank mRNA | AK010325.1 | - | 1 - 2 | 306 | 81 | |
AK030340.1 | - | 1 - 2 | 279 | 81 | ||
AK048012.1 | - | 1 - 2 | 268 | 81 | ||
AK050338.1 | - | 1 - 2 | 296 | 81 | ||
AK075720.1 | - | 1 - 2 | 349 | 81 | ||
AK088905.1 | - | 1 - 2 | 321 | 81 | ||
AK159757.1 | - | 1 - 2 | 294 | 81 | ||
BB619330.1 | - | 1 - 2 | 350 | 81 | ||
BC003862.1 | - | 1 - 2 | 288 | 3488326 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |