Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152817.4 | NP_690030.3 | 14 - 15 | 1949 | 123 | |
GenBank mRNA | AK012131.1 | - | 14 - 15 | 1949 | 123 | |
AK030942.1 | - | 14 - 15 | 1950 | 123 | ||
AK049324.1 | - | 14 - 15 | 1950 | 123 | ||
BC021912.1 | - | 13 - 14 | 1684 | 4503335 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |