Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020252.3 | NP_064648.3 | 21 - 22 | 5209 | 81 | |
NM_177284.2 | NP_796258.2 | 19 - 20 | 5173 | 81 | ||
GenBank mRNA | AB093249.2 | - | 21 - 22 | 5209 | 81 | |
AK017578.1 | - | 4 - 5 | 774 | 81 | ||
BC079893.1 | - | 19 - 20 | 5104 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |