Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027354.2 | NP_081630.2 | 5 - 6 | 944 | 87 | |
GenBank mRNA | AK011064.1 | - | 5 - 6 | 622 | 87 | |
AK154163.1 | - | 5 - 6 | 694 | 87 | ||
BC034901.1 | - | 5 - 6 | 604 | 5255069 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |