Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027354.2 | NP_081630.2 | 7 - 8 | 1189 | 63 | |
GenBank mRNA | AK011064.1 | - | 7 - 8 | 867 | 63 | |
AK081217.1 | - | 374 | 63 | |||
AK154163.1 | - | 7 - 8 | 939 | 63 | ||
BC034901.1 | - | 7 - 8 | 849 | 5255069 | 63 | |
BE981233.1 | - | 4 - 5 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |