Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_211138.1 | NP_997593.1 | 1 - 2 | 364 | 87 | |
GenBank mRNA | AK032027.1 | - | 1 - 2 | 66 | 87 | |
AY189949.1 | - | 1 - 2 | 201 | 87 | ||
BB646993.1 | - | 1 - 2 | 374 | 87 | ||
BC048917.1 | - | 1 - 2 | 235 | 6309383 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |