Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175639.1 | NP_783570.1 | 12 - 13 | 1570 | 65 | |
GenBank mRNA | AK012043.1 | - | 12 - 13 | 1555 | 65 | |
AK032907.1 | - | 12 - 13 | 1544 | 65 | ||
AK129031.1 | - | 12 - 13 | 1488 | 65 | ||
AK152321.1 | - | 12 - 13 | 1560 | 65 | ||
BC079913.1 | - | 11 - 12 | 1271 | 65 | ||
BC117724.2 | - | 13 - 14 | 1771 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |