Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021287.1 | NP_067262.1 | 27 - 28 | 6257 | 96 | |
GenBank mRNA | AK147671.1 | - | 27 - 28 | 6273 | 96 | |
AK172931.1 | - | 17 - 18 | 4566 | 96 | ||
BC048851.1 | - | 5 - 6 | 1075 | 96 | ||
BC079628.1 | - | 5 - 6 | 1209 | 96 | ||
BC079860.1 | - | 27 - 28 | 6257 | 6834745 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |