Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021287.1 | NP_067262.1 | 28 - 29 | 6394 | 61 | |
GenBank mRNA | AK147671.1 | - | 28 - 29 | 6410 | 61 | |
AK172931.1 | - | 18 - 19 | 4703 | 61 | ||
BC048851.1 | - | 6 - 7 | 1212 | 61 | ||
BC079628.1 | - | 6 - 7 | 1346 | 61 | ||
BC079860.1 | - | 28 - 29 | 6394 | 6834745 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |