Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167746.1 | NP_001161218.1 | 10 - 11 | 1555 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |