Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252573.1 | NP_001239502.1 | 6 - 7 | 916 | 70 | |
NM_001252574.1 | NP_001239503.1 | 6 - 7 | 851 | 70 | ||
NM_001252575.1 | NP_001239504.1 | 6 - 7 | 799 | 70 | ||
NM_144893.2 | NP_659142.1 | 6 - 7 | 870 | 70 | ||
NR_045544.1 | 7 - 8 | 747 | 70 | |||
GenBank mRNA | AK053180.1 | - | 6 - 7 | 786 | 70 | |
AK155539.1 | - | 6 - 7 | 885 | 70 | ||
AK160899.1 | - | 6 - 7 | 856 | 70 | ||
AK170715.1 | - | 6 - 7 | 821 | 70 | ||
AK189396.1 | - | 1 - 2 | 46 | 70 | ||
BC018327.1 | - | 7 - 8 | 736 | 4166249 | 70 | |
BC094025.1 | - | 6 - 7 | 849 | 4488690 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |