Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166685.1 | NP_001160157.1 | 9 - 10 | 1116 | 74 | |
NM_177088.3 | NP_796062.2 | 10 - 11 | 1245 | 74 | ||
GenBank mRNA | AK052705.1 | - | 10 - 11 | 1173 | 74 | |
AK076400.1 | - | 10 - 11 | 1223 | 74 | ||
BC090675.1 | - | 10 - 11 | 1201 | 74 | ||
BC116308.1 | - | 9 - 10 | 1082 | 40095242 | 74 | |
BC116309.1 | - | 9 - 10 | 1095 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |