Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010764.2 | NP_034894.2 | 8 - 9 | 1169 | 77 | |
GenBank mRNA | AK004817.2 | - | 8 - 9 | 1126 | 77 | |
AK035551.1 | - | 8 - 9 | 1124 | 77 | ||
AK147928.1 | - | 8 - 9 | 1127 | 77 | ||
AK150154.1 | - | 5 - 6 | 501 | 77 | ||
AK170848.1 | - | 5 - 6 | 500 | 77 | ||
AK171166.1 | - | 8 - 9 | 1127 | 77 | ||
AK171377.1 | - | 5 - 6 | 500 | 77 | ||
BC005430.1 | - | 8 - 9 | 1114 | 3488681 | 77 | |
U29947.1 | - | 8 - 9 | 1112 | 77 | ||
U87240.1 | - | 8 - 9 | 1103 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |