Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160414.1 | NP_001153886.1 | 3 - 4 | 346 | 83 | |
GenBank mRNA | AK136630.1 | - | 3 - 4 | 319 | 83 | |
AV381411.2 | - | 3 - 4 | 360 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons |