Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gse1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145896.1 | NP_001139368.1 | 8 - 9 | 1786 | 68 | |
NM_001145897.1 | NP_001139369.1 | 8 - 9 | 1730 | 68 | ||
NM_198671.2 | NP_941073.2 | 8 - 9 | 1918 | 68 | ||
GenBank mRNA | AK033263.1 | - | 8 - 9 | 1733 | 68 | |
AK042915.1 | - | 8 - 9 | 1735 | 68 | ||
AK122216.1 | - | 5 - 6 | 1068 | 68 | ||
AK154829.1 | - | 8 - 9 | 1765 | 68 | ||
BC043094.1 | - | 8 - 9 | 1918 | 6405464 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |