Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175561.4 | NP_780770.2 | 24 - 25 | 4315 | 61 | |
GenBank mRNA | AK087907.1 | - | 9 - 10 | 1228 | 61 | |
AK220342.1 | - | 16 - 17 | 1877 | 61 | ||
BC068235.1 | - | 13 - 14 | 2382 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |