Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011044.2 | NP_035174.1 | 3 - 4 | 551 | 78 | |
GenBank mRNA | AK028046.1 | - | 3 - 4 | 552 | 78 | |
AK030327.1 | - | 3 - 4 | 551 | 78 | ||
AK133496.1 | - | 3 - 4 | 552 | 78 | ||
AK149525.1 | - | 3 - 4 | 552 | 78 | ||
BC028342.1 | - | 3 - 4 | 553 | 78 | ||
BC037629.1 | - | 3 - 4 | 546 | 4973187 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |