Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172760.3 | NP_766348.3 | 16 - 17 | 1820 | 105 | |
GenBank mRNA | AK037067.1 | - | 16 - 17 | 1813 | 105 | |
BC018516.1 | - | 14 - 15 | 1709 | 3155913 | 105 | |
BC026617.1 | - | 11 - 12 | 1765 | 105 | ||
BC058752.1 | - | 16 - 17 | 1803 | 6485393 | 105 | |
BC138887.1 | - | 16 - 17 | 1777 | 8861909 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |