Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145607.3 | NP_663582.2 | 3 - 4 | 411 | 56 | |
GenBank mRNA | AK051200.1 | - | 3 - 4 | 418 | 56 | |
AK144842.1 | - | 3 - 4 | 412 | 56 | ||
AK148102.1 | - | 3 - 4 | 410 | 56 | ||
AK160894.1 | - | 3 - 4 | 420 | 56 | ||
BC072577.1 | - | 3 - 4 | 404 | 56 | ||
BC079851.1 | - | 3 - 4 | 400 | 6401930 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |